(2-Chlorphenyl)acetylen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Chloro-2-ethynylbenzene

M-Clarity™ Qualitätsstufe = MQ100

465305-1G
SIAL465305-1GEA 80.6 EUR
SIAL465305-1G
(2-Chlorphenyl)acetylen, Sigma-Aldrich®
(2-Chlorphenyl)acetylen
Formel: C₈H₅Cl
Molecular Weight: 136,58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00269951
CAS-Nummer: 873-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR