2,5-Bis(trifluormethyl)benzylbromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

465836-1G
SIAL465836-1GEA 54.8 EUR
SIAL465836-1G
2,5-Bis(trifluormethyl)benzylbromid, Sigma-Aldrich®
2,5-Bis(trifluormethyl)benzylbromid
Formel: C₉H₅BrF₆
MDL: MFCD00799485
CAS-Nummer: 302911-98-4
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR