(2,5-Bis(trifluormethyl)phenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

465852-1G
SIAL465852-1GEA 109 EUR
SIAL465852-1G
(2,5-Bis(trifluormethyl)phenyl)essigsäure, Sigma-Aldrich®
(2,5-Bis(trifluormethyl)phenyl)essigsäure
Formel: (C₆H₃(CF₃)₂)OCH₂COOH
Molecular Weight: 272,15 g/mol
CAS-Nummer: 302912-02-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR