4'-Fluor-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-2-hydroxyacetophenone

M-Clarity™ Qualitätsstufe = MQ100

466964-5G
SIAL466964-5GEA 38.8 EUR
SIAL466964-5G
4'-Fluor-2'-hydroxyacetophenon, Sigma-Aldrich®
4'-Fluor-2'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00143203
CAS-Nummer: 1481-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR