1-Fluor-2-ethinylbenzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Ethinyl-2-fluorbenzol, (2-Fluorphenyl)ethin, (2-Fluorphenyl)acetylen

467006-1G
SIAL467006-1GEA 33.6 EUR
SIAL467006-1G
1-Fluor-2-ethinylbenzol, Sigma-Aldrich®
1-Fluor-2-ethinylbenzol
Formel: C₈H₅F
Lagertemperatur: Kühlschrank
MDL: MFCD00799540
CAS-Nummer: 766-49-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR