2-Benzo[b]thiophencarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

467464-5G
SIAL467464-5GEA 76.5 EUR
SIAL467464-5G
2-Benzo[b]thiophencarbonsäure, Sigma-Aldrich®
2-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
Molecular Weight: 178,22 g/mol
MDL: MFCD00051636
CAS-Nummer: 6314-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR