3,4,5-Trichlorpyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

467936-1G
SIAL467936-1GEA 51.5 EUR
SIAL467936-1G
3,4,5-Trichlorpyridin, Sigma-Aldrich®
3,4,5-Trichlorpyridin
Formel: C₅H₂Cl₃N
Molecular Weight: 182,44 g/mol
MDL: MFCD00051685
CAS-Nummer: 33216-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR