(2-Bromethyl)cyclohexan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

467952-5G
SIAL467952-5GEA 72.2 EUR
SIAL467952-5G
(2-Bromethyl)cyclohexan, Sigma-Aldrich®
(2-Bromethyl)cyclohexan
Formel: C₈H₁₅Br
MDL: MFCD00019398
CAS-Nummer: 1647-26-3
EINECS: 216-712-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR