(R)-(+)-2-Bromisovaleriansäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-(+)-2-Bromo-3-methylbutyric acid, (R)-2-Bromoisovaleric acid, (R)-2-Bromo-3-methylbutyric acid
468347-5G
SIAL468347-5GEA
33.4
EUR
SIAL468347-5G
(R)-(+)-2-Bromisovaleriansäure, Sigma-Aldrich®
(R)-(+)-2-Bromisovaleriansäure
Formel:
C₅H₉BrO₂ Molecular Weight: 181,03 g/mol |
MDL:
MFCD00210113 CAS-Nummer: 76792-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...