4,5-Difluor-2-nitrobenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

468983-5G
SIAL468983-5GEA 99.9 EUR
SIAL468983-5G
4,5-Difluor-2-nitrobenzoesäure, Sigma-Aldrich®
4,5-Difluor-2-nitrobenzoesäure
Formel: C₇H₃F₂NO₄
Molecular Weight: 203,1 g/mol
MDL: MFCD00799521
CAS-Nummer: 20372-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR