1-Pentylmagnesiumchlorid 2 mol/l in THF, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pentylmagnesium chloride

M-Clarity™ Qualitätsstufe = MQ100

469033-100G
SIAL469033-100GEA 226 EUR
SIAL469033-100G
1-Pentylmagnesiumchlorid 2 mol/l in THF, Sigma-Aldrich®
1-Pentylmagnesiumchlorid
Formel: C₅H₁₁ClMg
Molecular Weight: 130,9 g/mol
MDL: MFCD00672089
CAS-Nummer: 6393-56-2
UN: 2924
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR