Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

46920-100G-F 46920-25G-F 46920-5G-F
SIAL46920-100G-FEA 355 EUR
SIAL46920-100G-F SIAL46920-25G-F SIAL46920-5G-F
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid), Sigma-Aldrich®
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel: C₁₉H₁₅NO₅
MDL: MFCD00010733
CAS-Nummer: 82911-69-1
EINECS: 433-520-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR