Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
46920-100G-F
46920-25G-F
46920-5G-F
SIAL46920-100G-FEA
355
EUR
SIAL46920-100G-F
SIAL46920-25G-F
SIAL46920-5G-F
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid), Sigma-Aldrich®
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel:
C₁₉H₁₅NO₅ |
MDL:
MFCD00010733 CAS-Nummer: 82911-69-1 EINECS: 433-520-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...