N-Boc-D-alaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-D-Alaninol, (R)-2-(BOC-amino)-1-propanol

M-Clarity™ Qualitätsstufe = MQ100

469505-5G
SIAL469505-5GEA 148 EUR
SIAL469505-5G
N-Boc-D-alaninol, Sigma-Aldrich®
N-Boc-D-alaninol
Formel: C₈H₁₇NO₃
Molecular Weight: 175,2 g/mol
MDL: MFCD00235912
CAS-Nummer: 106391-86-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR