N-Boc-L-alaninol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-L-Alaninol, Boc-Alaninol, (S)-2-(BOC-amino)-1-propanol

M-Clarity™ Qualitätsstufe = MQ100

469513-5G
SIAL469513-5GEA 142 EUR
SIAL469513-5G
N-Boc-L-alaninol, Sigma-Aldrich®
N-Boc-L-alaninol
Formel: C₈H₁₇NO₃
Molecular Weight: 175,2 g/mol
MDL: MFCD00043121
CAS-Nummer: 79069-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR