3-Methoxypyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

469890-5G
SIAL469890-5GEA 101 EUR
SIAL469890-5G
3-Methoxypyridin, Sigma-Aldrich®
3-Methoxypyridine
Formel: C₆H₇NO
Molecular Weight: 109,13 g/mol
MDL: MFCD00673022
CAS-Nummer: 7295-76-3
EINECS: 230-730-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR