4-(Trifluormethylthio)phenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-[(Trifluoromethyl)sulphanyl]phenol

M-Clarity™ Qualitätsstufe = MQ100

470082-5G
SIAL470082-5GEA 24.6 EUR
SIAL470082-5G
4-(Trifluormethylthio)phenol, Sigma-Aldrich®
4-(Trifluormethylthio)phenol
Formel: C₇H₅F₃OS
Molecular Weight: 194,18 g/mol
MDL: MFCD00036035
CAS-Nummer: 461-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR