4-(Difluormethoxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

470112-5G
SIAL470112-5GEA 45.6 EUR
SIAL470112-5G
4-(Difluormethoxy)benzaldehyd, Sigma-Aldrich®
4-(Difluormethoxy)benzaldehyd
Formel: C₈H₆F₂O₂
MDL: MFCD00042252
CAS-Nummer: 73960-07-3
EINECS: 277-653-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR