2-(Propylamino)ethanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
N-Propylethanolamine
M-Clarity™ Qualitätsstufe = MQ100
470201-25ML
SIAL470201-25MLEA
170
EUR
SIAL470201-25ML
2-(Propylamino)ethanol, Sigma-Aldrich®
2-(Propylamino)ethanol
Formel:
C₅H₁₃NO Molecular Weight: 103,16 g/mol Siedepunkt: 182 °C (1013 hPa) Dichte: 0,901 g/cm³ (20 °C) |
MDL:
MFCD00002844 CAS-Nummer: 16369-21-4 EINECS: 240-426-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...