α,α,α-Trifluor-m-xylol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-(Trifluormethyl)toluol, 3-Methylbenzotrifluorid, 1-Methyl-3-(trifluormethyl)benzol
M-Clarity™ Qualitätsstufe = MQ100
470384-5G
SIAL470384-5GEA
98.9
EUR
SIAL470384-5G
α,α,α-Trifluor-m-xylol, Sigma-Aldrich®
α,α,α-Trifluor-m-xylol
Formel:
C₈H₇F₃ Molecular Weight: 160,14 g/mol Siedepunkt: 127 °C (1013 hPa) Dichte: 1,148 g/cm³ (20 °C) |
MDL:
MFCD00190105 CAS-Nummer: 401-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...