Methyl-2,6-dimethoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dimethoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

470694-1G
SIAL470694-1GEA 39 EUR
SIAL470694-1G
Methyl-2,6-dimethoxybenzoat, Sigma-Aldrich®
Methyl-2,6-dimethoxybenzoat
Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
MDL: MFCD00075861
CAS-Nummer: 2065-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR