Antimon(III)isopropoxid, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

470732-5ML
SIAL470732-5MLEA 220 EUR
SIAL470732-5ML
Antimon(III)isopropoxid, Sigma-Aldrich®
Antimon(III)isopropoxid
Formel: C₉H₂₁O₃Sb
Molecular Weight: 298,80 g/mol
MDL: MFCD00143623
CAS-Nummer: 18770-47-3
EINECS: 242-561-6
UN: 3082
ADR: 9,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR