4-Acetylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

M-Clarity™ Qualitätsstufe = MQ100

470821-25G 470821-5G
SIAL470821-25GEA 284 EUR
SIAL470821-25G SIAL470821-5G
4-Acetylphenylboronsäure, Sigma-Aldrich®
4-Acetylphenylboronsäure
Formel: C₈H₉BO₃
Molecular Weight: 163.97
MDL: MFCD01074667
CAS-Nummer: 149104-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR