3,5-Bis(trifluormethyl)phenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3,5-Bis(trifluormethyl)phenyl)metaborsäure, 3,5-Bis(trifluormethyl)benzolboronsäure, Dihydroxy(3,5-bis(trifluormethyl)phenyl)boran

M-Clarity™ Qualitätsstufe = MQ100

471070-5G
SIAL471070-5GEA 94.2 EUR
SIAL471070-5G
3,5-Bis(trifluormethyl)phenylboronsäure, Sigma-Aldrich®
3,5-Bis(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₆O₂
MDL: MFCD00051850
CAS-Nummer: 73852-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR