1-(3-Methoxyphenyl)piperazin, Sigma-Aldrich®
Lieferant: Merck
471682-5G
SIAL471682-5GEA
29.8
EUR
SIAL471682-5G
1-(3-Methoxyphenyl)piperazin, Sigma-Aldrich®
1-(3-Methoxyphenyl)piperazin
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,26 g/mol Siedepunkt: 130 °C (0,5 mmHg) Dichte: 1,082 g/cm³ (20 °C) |
MDL:
MFCD00040733 CAS-Nummer: 16015-71-7 EINECS: 240-154-8 UN: 3267 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...