(S)-(+)-Ketoprofen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-(3-Benzoylphenyl)propanoic acid, S-(+)-Ketoprofen

M-Clarity™ Qualitätsstufe = MQ100

471909-1G
SIAL471909-1GEA 134 EUR
SIAL471909-1G
(S)-(+)-Ketoprofen, Sigma-Aldrich®
(S)-(+)-Ketoprofen
Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
MDL: MFCD00673316
CAS-Nummer: 22161-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR