9-Oxabicyclo[6.1.0]-4-nonen, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

471984-5G
SIAL471984-5GEA 114 EUR
SIAL471984-5G
9-Oxabicyclo[6.1.0]-4-nonen, Sigma-Aldrich®
9-Oxabicyclo[6.1.0]-4-nonen
Formel: C₈H₁₂O
Molecular Weight: 124,18 g/mol
MDL: MFCD00086420
CAS-Nummer: 637-90-1
EINECS: 211-308-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR