1-Chlor-2-methylbutan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

473324-50ML
SIAL473324-50MLEA 113 EUR
SIAL473324-50ML
1-Chlor-2-methylbutan, Sigma-Aldrich®
1-Chlor-2-methylbutan
Formel: C₅H₁₁Cl
Molecular Weight: 106,6 g/mol
MDL: MFCD00039366
CAS-Nummer: 616-13-7
EINECS: 210-466-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR