7-Chlorindol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

473731-1G
SIAL473731-1GEA 125 EUR
SIAL473731-1G
7-Chlorindol, Sigma-Aldrich®
7-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151,6 g/mol
MDL: MFCD01321301
CAS-Nummer: 53924-05-3
EINECS: 258-865-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR