Fmoc-D-phenylalanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fmoc-D-Phe-OH, N-Fmoc-D-phenylalanine

47378-5G
SIAL47378-5GEA 95.3 EUR
SIAL47378-5G
Fmoc-D-phenylalanin, Sigma-Aldrich®
Fmoc-D-phenylalanin
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387.4 g/mol
MDL: MFCD00062955
CAS-Nummer: 86123-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR