1-Iod-3,3,3-trifluorpropan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

473820-1G 473820-5G
SIAL473820-1GEA 22.8 EUR
SIAL473820-1G SIAL473820-5G
1-Iod-3,3,3-trifluorpropan, Sigma-Aldrich®
1-Iod-3,3,3-trifluorpropan
Formel: C₃H₄F₃I
MDL: MFCD00038531
CAS-Nummer: 460-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR