(S)-(-)-3-Amino-1,2-propandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-(-)-3-Amino-1,2-dihydroxypropan

M-Clarity™ Qualitätsstufe = MQ200

473960-1G
SIAL473960-1GEA 32.3 EUR
SIAL473960-1G
(S)-(-)-3-Amino-1,2-propandiol, Sigma-Aldrich®
(S)-(-)-3-Amino-1,2-propandiol
Formel: C₃H₉NO₂
Molecular Weight: 91,11 g/mol
MDL: MFCD00798260
CAS-Nummer: 61278-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR