(R)-(+)-2-(4-Hydroxyphenoxy)propionsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-2-(4-Hydroxyphenoxy)propanoic acid
M-Clarity™ Qualitätsstufe = MQ100
474533-25G
SIAL474533-25GEA
40
EUR
SIAL474533-25G
(R)-(+)-2-(4-Hydroxyphenoxy)propionsäure, Sigma-Aldrich®
(R)-(+)-2-(4-Hydroxyphenoxy)propionsäure
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol |
MDL:
MFCD00274088 CAS-Nummer: 94050-90-5 EINECS: 407-960-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...