N-Fmoc-D-valin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fmoc-D-Val-OH, D-Valine, N-FMOC protected

47481-5G
SIAL47481-5GEA 74.6 EUR
SIAL47481-5G
N-Fmoc-D-valin, Sigma-Aldrich®
N-Fmoc-D-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339,39 g/mol
MDL: MFCD00062953
CAS-Nummer: 84624-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR