Fmoc-D-Pro-OH, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Fmoc-D-proline

47532-5G
SIAL47532-5GEA 123 EUR
SIAL47532-5G
Fmoc-D-Pro-OH, Sigma-Aldrich®
Fmoc-D-Pro-OH
Formel: C₂₀H₁₉NO₄
Molecular Weight: 337,38 g/mol
MDL: MFCD00077067
CAS-Nummer: 101555-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR