Monoethylsuccinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl hydrogen succinate, 4-Ethoxy-4-oxobutanoic acid, mono-Ethyl succinate

M-Clarity™ Qualitätsstufe = MQ100

475505-5G
SIAL475505-5GEA 83.8 EUR
SIAL475505-5G
Monoethylsuccinat, Sigma-Aldrich®
Monoethylsuccinat
Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
MDL: MFCD00053346
CAS-Nummer: 1070-34-4
EINECS: 213-973-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR