Polyethylene glycol diacrylate (PEGDA) ≥92,0% Mn 250 inhibiert, Sigma-Aldrich®

Lieferant: Merck
Gefahr


Stabilisierung: Stabilisiert mit 4-Methoxyphenol (MEHQ) 100 ppm
M-Clarity™ Qualitätsstufe = MQ100

475629-100ML 475629-500ML
SIAL475629-100MLEA 72 EUR
SIAL475629-100ML SIAL475629-500ML
Polyethylene glycol diacrylate (PEGDA) ≥92,0% Mn 250 inhibiert, Sigma-Aldrich®
Polyethylene glycol diacrylate (PEGDA)
Formel: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Dichte: 1,11 g/ml (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00081876
CAS-Nummer: 26570-48-9

Bestellen

Spezifikation Testergebnisse

Refractive Index n20/D 1.463
Ω-end Acrylate
α-end Acrylate
Water (by Karl Fischer) ≤1.00%
Acid value ≤1.00 (mg KOH/g)
Color test ≤200 APHA
Inhibitor (MEHQ) 80 - 120 ppm
Purity ≥92.0%
Proton NMR Spectra Conforms to Structure
Polymer Architecture (Shape) Linear
Polymer Architecture (Functionality) Homobifunctional
Reaction Suitability (Reagent Type) Cross-linking Reagent
Reaction Suitability (Reaction Type) Polymerisation Reactions


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR