N-alpha-FMOC-Nepsilon-BOC-L-Lysin ≥98,0% (durch HPLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-ε-(tert-Butoxycarbonyl)-N-ɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-((tert-butoxycarbonyl)amino)hexanoic acid, N-ε-Boc-N-ɑ-Fmoc-L-lysine, Fmoc-Lys(Boc)-OH

47624-1G-F 47624-10G-F 47624-25G-F 47624-100G-F 47624-1KG-F
SIAL47624-1G-FEA 24.8 EUR
SIAL47624-1G-F SIAL47624-10G-F SIAL47624-25G-F SIAL47624-100G-F SIAL47624-1KG-F
N-alpha-FMOC-Nepsilon-BOC-L-Lysin ≥98,0% (durch HPLC), Sigma-Aldrich®
N-alpha-FMOC-Nepsilon-BOC-L-Lysin
Formel: C₂₆H₃₂N₂O₆
Lagertemperatur: Kühlschrank
MDL: MFCD00037138
CAS-Nummer: 71989-26-9

Bestellen

Spezifikation Testergebnisse

Proton NMR Spectrum Conforms to Structure
Specific Rotation (C=1 in DMF at 20 °C) -13 to -11°
Purity (HPLC) ≥98.0%
Optical Activity [α]20/D - 12±1°, c = 1% in DMF


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR