3,4'-Diaminodiphenylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(4-Aminophenoxy)aniline, 3,4'-Oxydianiline

M-Clarity™ Qualitätsstufe = MQ100

476307-10G
SIAL476307-10GEA 118 EUR
SIAL476307-10G
3,4'-Diaminodiphenylether, Sigma-Aldrich®
3,4'-Diaminodiphenylether
Formel: C₁₂H₁₂N₂O
Molecular Weight: 200,2365 g/mol
MDL: MFCD00036097
CAS-Nummer: 2657-87-6
EINECS: 220-190-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR