1,3-Bis(4-aminophenoxy)benzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

476323-100G
SIAL476323-100GEA 248 EUR
SIAL476323-100G
1,3-Bis(4-aminophenoxy)benzol, Sigma-Aldrich®
1,3-Bis(4-aminophenoxy)benzol
Formel: C₁₈H₁₆N₂O₂
Molecular Weight: 292,3300 g/mol
MDL: MFCD00039154
CAS-Nummer: 2479-46-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR