N-Fmoc-L-valin, Sigma-Aldrich®

Lieferant: Merck

47638-50G-F
SIAL47638-50G-FEA 74.3 EUR
SIAL47638-50G-F
N-Fmoc-L-valin, Sigma-Aldrich®
N-Fmoc-L-valin
Formel: C₂₀H₂₁NO₄
Molecular Weight: 339.4 g/mol
MDL: MFCD00037124
CAS-Nummer: 68858-20-8
EINECS: 272-515-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR