2-Methylbutyrylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DL-2-Methylbutyryl chloride

M-Clarity™ Qualitätsstufe = MQ100

476471-5G
SIAL476471-5GEA 65 EUR
SIAL476471-5G
2-Methylbutyrylchlorid, Sigma-Aldrich®
2-Methylbutyrylchlorid
Formel: C₅H₉ClO
Molecular Weight: 120,58 g/mol
MDL: MFCD00041726
CAS-Nummer: 57526-28-0
EINECS: 260-787-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR