5-Bromisatin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Brom-2,3-indolindion, 5-Brom-1H-indol-2,3-dion

M-Clarity™ Qualitätsstufe = MQ100

476994-25G 476994-5G
SIAL476994-25GEA 65.3 EUR
SIAL476994-25G SIAL476994-5G
5-Bromisatin, Sigma-Aldrich®
5-Bromisatin
Formel: C₈H₄BrNO₂
Molecular Weight: 226.03 g/mol
MDL: MFCD00149345
CAS-Nummer: 87-48-9
EINECS: 201-747-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR