(Formylmethyl)triphenylphosphoniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Oxoethyl)triphenylphosphonium chloride

M-Clarity™ Qualitätsstufe = MQ100

47718-10G
SIAL47718-10GEA 65.8 EUR
SIAL47718-10G
(Formylmethyl)triphenylphosphoniumchlorid, Sigma-Aldrich®
(Formylmethyl)triphenylphosphoniumchlorid
Formel: C₂₀H₁₈ClOP
MDL: MFCD00012003
CAS-Nummer: 62942-43-2
EINECS: 263-767-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR