5-tert-Butylsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-(tert-Butyl)-2-hydroxybenzaldehyde, 5-tert-Butyl-2-hydroxybenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

477532-1G
SIAL477532-1GEA 107 EUR
SIAL477532-1G
5-tert-Butylsalicylaldehyd, Sigma-Aldrich®
5-tert-Butylsalicylaldehyd
Formel: C₁₁H₁₄O₂
Molecular Weight: 178,23 g/mol
MDL: MFCD01321335
CAS-Nummer: 2725-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR