3,5-Dimethoxyphenylisocyanat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
478164-2G
SIAL478164-2GEA
147
EUR
SIAL478164-2G
3,5-Dimethoxyphenylisocyanat, Sigma-Aldrich®
3,5-Dimethoxyphenylisocyanat
Formel:
C₉H₉NO₃ Molecular Weight: 179,18 g/mol Lagertemperatur: Kühlschrank |
CAS-Nummer:
54132-76-2 UN: 3335 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...