Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), Zr(TMHD)4

M-Clarity™ Qualitätsstufe = MQ100

478865-5G
SIAL478865-5GEA 122 EUR
SIAL478865-5G
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV), Sigma-Aldrich®
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')zirkonium(IV)
Formel: C₄₄H₇₆O₈Zr
Molecular Weight: 824,29 g/mol
MDL: MFCD00145380
CAS-Nummer: 18865-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR