4-Brombenzaldehyddiethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Brom-α,α-diethoxytoluol, 1-Brom-4-(diethoxymethyl)benzol

M-Clarity™ Qualitätsstufe = MQ100

478962-10ML
SIAL478962-10MLEA 58.3 EUR
SIAL478962-10ML
4-Brombenzaldehyddiethylacetal, Sigma-Aldrich®
4-Brombenzaldehyddiethylacetal
Formel: C₁₁H₁₅BrO₂
Molecular Weight: 259,14 g/mol
MDL: MFCD01863514
CAS-Nummer: 34421-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR