3-Acetoxybenzoesäure, Sigma-Aldrich®

Lieferant: Merck

479349-10G
SIAL479349-10GEA 64.3 EUR
SIAL479349-10G
3-Acetoxybenzoesäure, Sigma-Aldrich®
3-Acetoxybenzoesäure
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00016699
CAS-Nummer: 6304-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR