1,2-Bis(dicyclohexylphosphino)ethan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

479500-1G 479500-5G
SIAL479500-1GEA 116 EUR
SIAL479500-1G SIAL479500-5G
1,2-Bis(dicyclohexylphosphino)ethan, Sigma-Aldrich®
1,2-Bis(dicyclohexylphosphino)ethan
Formel: C₂₆H₄₈P₂
MDL: MFCD00015521
CAS-Nummer: 23743-26-2
UN: 1325
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR